missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Respiratory Syncytial Virus (RSV) A2 Fusion Protein Rabbit anti-Virus, Clone: R338, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89761-200UG
This item is not returnable.
View return policy
Description
Human Respiratory Syncytial Virus (RSV) A2 Fusion Protein Monoclonal specifically detects Human Respiratory Syncytial Virus (RSV) A2 Fusion Protein in Virus samples. It is validated for Sandwich ELISA Capture, Block/Neutralize.
Specifications
| Human Respiratory Syncytial Virus (RSV) A2 Fusion Protein | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
| ELISA | |
| R338 | |
| Sandwich ELISA Capture 0.5-4 ug/ml | |
| HRSVgp08, RSV, RSV fusion protein | |
| Recombinant Human Respiratory Syncytial Virus (RSV) A2 Fusion Protein protein | |
| 200 μg | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction