missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human RELT/TNFRSF19L Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
£167.00 - £491.00
Specifications
| Antigen | RELT/TNFRSF19L |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18785277
![]() |
R&D Systems
AF1385-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18000762
![]() |
R&D Systems
AF1385 |
100 μg |
£491.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RELT/TNFRSF19L Polyclonal specifically detects RELT/TNFRSF19L in Human samples. It is validated for Western Blot.Specifications
| RELT/TNFRSF19L | |
| Western Blot 1 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| Receptor expressed in lymphoid tissues, RELT tumor necrosis factor receptor, TNFRSF19L, TNFRSF19LFLJ14993, tumor necrosis factor receptor superfamily member 19L, tumor necrosis factor receptor superfamily, member 19-like | |
| RELT | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q969Z4 | |
| 84957 | |
| Mouse myeloma cell line NS0-derived recombinant human RELT/TNFRSF19L Ser26-Ala160 (Arg127Gly, Arg129Gly) Accession # Q969Z4 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human RELT/TNFRSF19L in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) 4-1BB, rhBAFF R, rhCD27, rhTAJ, rhCD30, rhDR3, rhDR6, rhTNF RI, rhTNF RII, rhEDAR, rhFas, rhGITR, rhHVEM, rhNGF R, rhOPG, and recombinant mouse OX40 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title