Learn More
Human RELT/TNFRSF19L Antibody, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody has been used in 2 publications
Brand:  R&D Systems AF1385-SP
 View more versions of this product
Product Code. 18785277
  • £167.00 / 25µg
Description
RELT/TNFRSF19L Polyclonal specifically detects RELT/TNFRSF19L in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RELT/TNFRSF19L
Polyclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Receptor expressed in lymphoid tissues, RELT tumor necrosis factor receptor, TNFRSF19L, TNFRSF19LFLJ14993, tumor necrosis factor receptor superfamily member 19L, tumor necrosis factor receptor superfamily, member 19-like
Goat
Affinity Purified
RUO
84957
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Western Blot
LYOPH
Western Blot 1 ug/mL
Q969Z4
RELT
Mouse myeloma cell line NS0-derived recombinant human RELT/TNFRSF19L Ser26-Ala160 (Arg127Gly, Arg129Gly) Accession # Q969Z4
25 μg
Primary
Detects human RELT/TNFRSF19L in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) 4-1BB, rhBAFF R, rhCD27, rhTAJ, rhCD30, rhDR3, rhDR6, rhTNF RI, rhTNF RII, rhEDAR, rhFas, rhGITR, rhHVEM, rhNGF R, rhOPG, and recombinant mouse OX40 is observed.