missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human RELM beta Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
£157.00 - £352.00
Specifications
| Antigen | RELM beta |
|---|---|
| Clone | 250911 |
| Concentration | LYOPH |
| Dilution | ELISA |
| Applications | ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18748749
![]() |
R&D Systems
MAB2917-SP |
25 μg |
£157.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18168740
![]() |
R&D Systems
MAB2917 |
100 μg |
£352.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RELM beta Monoclonal specifically detects RELM beta in Human samples. It is validated for ELISA.Specifications
| RELM beta | |
| LYOPH | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9BQ08 | |
| 84666 | |
| E. coli-derived recombinant human RELM beta Gly21-Thr111 Accession # Q9BQ08 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human RELM beta in direct ELISAs. Does not cross-react with recombinant mouse (rm) RELM alpha , rmRELM beta , recombinant rat RELM gamma , or rmResistin. Not recommended for natural lysates in Western blot. |
| 250911 | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C/EBP-epsilon regulated myeloid-specific secreted cysteine-rich proteinprecursor 2, CCRG, Colon and small intestine-specific cysteine-rich protein, Colon carcinoma-related gene protein, cysteine-rich secreted A12-alpha-like protein 1, Cysteine-rich secreted protein A12-alpha-like 1, Cysteine-rich secreted protein FIZZ2, FIZZ1, FIZZ2RELM-beta, found in inflammatory zone 1, HXCP2XCP2, RELMb, RELMbeta, resistin like beta, resistin-like beta, RETNL2 | |
| RETNLB | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title