missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human RELM beta Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB2917
This item is not returnable.
View return policy
Description
RELM beta Monoclonal specifically detects RELM beta in Human samples. It is validated for ELISA.
Specifications
| RELM beta | |
| Monoclonal | |
| LYOPH | |
| ELISA | |
| Q9BQ08 | |
| RETNLB | |
| E. coli-derived recombinant human RELM beta Gly21-Thr111 Accession # Q9BQ08 | |
| 100 μg | |
| Primary | |
| Detects human RELM beta in direct ELISAs. Does not cross-react with recombinant mouse (rm) RELM alpha , rmRELM beta , recombinant rat RELM gamma , or rmResistin. Not recommended for natural lysates in Western blot. | |
| Human | |
| Purified |
| ELISA | |
| 250911 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C/EBP-epsilon regulated myeloid-specific secreted cysteine-rich proteinprecursor 2, CCRG, Colon and small intestine-specific cysteine-rich protein, Colon carcinoma-related gene protein, cysteine-rich secreted A12-alpha-like protein 1, Cysteine-rich secreted protein A12-alpha-like 1, Cysteine-rich secreted protein FIZZ2, FIZZ1, FIZZ2RELM-beta, found in inflammatory zone 1, HXCP2XCP2, RELMb, RELMbeta, resistin like beta, resistin-like beta, RETNL2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 84666 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction