missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Reg1B Antibody, R&D Systems™

Goat Polyclonal Antibody
£146.00 - £413.00
Specifications
| Antigen | Reg1B |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18777917
![]() |
R&D Systems
AF2090-SP |
25 μg |
£146.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18083102
|
R&D Systems
AF2090 |
100 μg |
£413.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Reg1B Polyclonal specifically detects Reg1B in Human samples. It is validated for Western Blot.Specifications
| Reg1B | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| Lithostathine 1 beta, lithostathine-1-beta, Pancreatic stone protein 2, pancreatic threadprotein), PSPS2, PSPS2PSP-2, regenerating islet-derived 1 beta, Regenerating islet-derived protein 1-beta, Regenerating protein I beta, REGH, REGLREG-1-beta, secretory pancreatic stone protein 2 | |
| REG1B | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| P48304 | |
| 5968 | |
| E. coli-derived recombinant human Reg1B Gln23-Asn166 Accession # P48304 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human Reg1B in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant mouse (rm) Reg1 is observed, 5% cross-reactivity with rmReg2 and recombinant rat Reg1 is observed and less than 1% cross-reactivity with recombinant human Reg4 and rmReg3A is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title