missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Reg1B Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF2090
This item is not returnable.
View return policy
Description
Reg1B Polyclonal specifically detects Reg1B in Human samples. It is validated for Western Blot.
Specifications
| Reg1B | |
| Polyclonal | |
| Unconjugated | |
| P48304 | |
| REG1B | |
| E. coli-derived recombinant human Reg1B Gln23-Asn166 Accession # P48304 | |
| 100 μg | |
| Primary | |
| Detects human Reg1B in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant mouse (rm) Reg1 is observed, 5% cross-reactivity with rmReg2 and recombinant rat Reg1 is observed and less than 1% cross-reactivity with recombinant human Reg4 and rmReg3A is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Lithostathine 1 beta, lithostathine-1-beta, Pancreatic stone protein 2, pancreatic threadprotein), PSPS2, PSPS2PSP-2, regenerating islet-derived 1 beta, Regenerating islet-derived protein 1-beta, Regenerating protein I beta, REGH, REGLREG-1-beta, secretory pancreatic stone protein 2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5968 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion