missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human RAGE Quantikine ELISA Kit

Product Code. 18741954
Change view
Click to view available options
Quantity:
1 Plate
6 Plates
Sufficient For:
1 x 96-well Plate
6 x 96-well Plates
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Sufficient For unitSize
18741954 1 Plate 1 x 96-well Plate 1 set
18722244 6 Plates 6 x 96-well Plates 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18741954 Supplier R&D Systems™ Supplier No. DRG00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human RAGE Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human RAGE Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human AGER in cell culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 78.00 - 5000pg/mL
Assay Sensitivity 16.14pg/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum
Specificity Natural and Recombinant Human RAGE
Sufficient For 1 x 96-well Plate
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 177
Quantity 1 Plate
Sample Volume Cell Culture Supernates (50uL),Serum (50uL),EDTA Plasma (50uL),Heparin Plasma (50uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym advanced glycosylation end product-specific receptor, AGER, RAGE isoform delta, RAGE isoform sRAGE-delta, Receptor for advanced glycosylation end products, receptor for advanced glycosylation end-products, SCARJ1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.