missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Pro Meprin beta Subunit/MEP1B Antibody, R&D Systems™

Rat Monoclonal Antibody
£146.00 - £345.00
Specifications
| Antigen | Meprin beta Subunit/MEP1B |
|---|---|
| Clone | 289703 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18791627
![]() |
R&D Systems
MAB28952-SP |
25 μg |
£146.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18131451
![]() |
R&D Systems
MAB28952 |
100 μg |
£345.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Meprin beta Subunit/MEP1B Monoclonal specifically detects Meprin beta Subunit/MEP1B in Human samples. It is validated for Western Blot.Specifications
| Meprin beta Subunit/MEP1B | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Human | |
| Q16820 | |
| 4225 | |
| Human embryonic kidney cell line HEK293-derived recombinant human Meprin beta Subunit/MEP1B Thr23-Ser593 Accession # Q16820 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human or mouse Pro Meprin beta Subunit/MEP1B in Western blots. It does not react with the mature form. |
| 289703 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.4.24.63, endopeptidase-2, MEP1B, meprin A subunit beta, meprin A, beta, Meprin B, N-benzoyl-L-tyrosyl-p-amino-benzoic acid hydrolase beta subunit, N-benzoyl-L-tyrosyl-P-amino-benzoic acid hydrolase subunit beta, PABA peptide hydrolase, PPH beta | |
| MEP1B | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title