missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Pro Meprin beta Subunit/MEP1B Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB28952-SP
This item is not returnable.
View return policy
Description
Meprin beta Subunit/MEP1B Monoclonal specifically detects Meprin beta Subunit/MEP1B in Human samples. It is validated for Western Blot.
Specifications
| Meprin beta Subunit/MEP1B | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q16820 | |
| MEP1B | |
| Human embryonic kidney cell line HEK293-derived recombinant human Meprin beta Subunit/MEP1B Thr23-Ser593 Accession # Q16820 | |
| 25 μg | |
| Primary | |
| Detects human or mouse Pro Meprin beta Subunit/MEP1B in Western blots. It does not react with the mature form. | |
| Human | |
| Purified |
| Western Blot | |
| 289703 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.4.24.63, endopeptidase-2, MEP1B, meprin A subunit beta, meprin A, beta, Meprin B, N-benzoyl-L-tyrosyl-p-amino-benzoic acid hydrolase beta subunit, N-benzoyl-L-tyrosyl-P-amino-benzoic acid hydrolase subunit beta, PABA peptide hydrolase, PPH beta | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4225 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction