missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Primate IL-15 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 11 publications
£184.00 - £1830.00
Specifications
| Antigen | IL-15 |
|---|---|
| Clone | 34505 |
| Dilution | Neutralization 0.1-0.2 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Neutralization, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18774588
![]() |
R&D Systems
MAB647-SP |
25 μg |
£184.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18159620
|
R&D Systems
MAB647-100 |
100 μg |
£467.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18778069
![]() |
R&D Systems
MAB647-500 |
500 μg |
£1830.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-15 Monoclonal specifically detects IL-15 in Human, Primate samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
| IL-15 | |
| Neutralization 0.1-0.2 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human, Primate | |
| P40933 | |
| 3600 | |
| E. coli-derived recombinant human IL-15 Asn49-Ser162 Accession # P40933 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human IL-15 in direct ELISAs and Western blots. In ELISAs, no cross-reactivity with recombinant human (rh) IL-1 alpha , -1 beta , -2, -3, -4, -5, -6, -7, -8, -9, rmIL-1 alpha , -1 beta , -3, -4, -5, -6, -7, -9, or -13 is observed. |
| 34505 | |
| Neutralization, ELISA | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| IL15, IL-15MGC9721, interleukin 15, interleukin-15 | |
| IL15 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title