missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Primate IL-15 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 11 publications
Brand: R&D Systems MAB647-SP
This item is not returnable.
View return policy
Description
IL-15 Monoclonal specifically detects IL-15 in Human, Primate samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications
| IL-15 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| IL15, IL-15MGC9721, interleukin 15, interleukin-15 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3600 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Neutralization, ELISA | |
| 34505 | |
| Neutralization 0.1-0.2 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P40933 | |
| IL15 | |
| E. coli-derived recombinant human IL-15 Asn49-Ser162 Accession # P40933 | |
| 25 μg | |
| Primary | |
| Detects human IL-15 in direct ELISAs and Western blots. In ELISAs, no cross-reactivity with recombinant human (rh) IL-1 alpha , -1 beta , -2, -3, -4, -5, -6, -7, -8, -9, rmIL-1 alpha , -1 beta , -3, -4, -5, -6, -7, -9, or -13 is observed. | |
| Human, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction