missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Presenilin-1 N-Terminal Fragment Antibody, R&D Systems™

Mouse Monoclonal Antibody
£156.00 - £452.00
Specifications
| Antigen | Presenilin-1 |
|---|---|
| Clone | 121130 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18724279
![]() |
R&D Systems
MAB1491-SP |
25 μg |
£156.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18159040
![]() |
R&D Systems
MAB1491 |
500 μg |
£452.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Presenilin-1 Monoclonal specifically detects Presenilin-1 in Human samples. It is validated for Western Blot.Specifications
| Presenilin-1 | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P49768 | |
| 5663 | |
| E. coli-derived recombinant human Presenilin-1 N-Terminal Fragment Met1-Lys80 Accession # P49768 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Presenilin-1 N-Terminal Fragment in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) Presenilin-1 C-Terminal Fragment, rhPresenilin-2 C-Terminal Fragment, or rhPresenilin-2 N-Terminal Fragment is observed. |
| 121130 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AD3EC 3.4.23.-, Alzheimer disease 3, EC 3.4.23, FAD, presenilin 1, Presenilin1, Protein S182, PS-1, PS1presenilin-1, PSEN1, PSNL1, S182 | |
| PSEN1 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title