missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Presenilin-1 N-Terminal Fragment Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB1491-SP
This item is not returnable.
View return policy
Description
Presenilin-1 Monoclonal specifically detects Presenilin-1 in Human samples. It is validated for Western Blot.
Specifications
| Presenilin-1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P49768 | |
| PSEN1 | |
| E. coli-derived recombinant human Presenilin-1 N-Terminal Fragment Met1-Lys80 Accession # P49768 | |
| 25 μg | |
| Primary | |
| Detects human Presenilin-1 N-Terminal Fragment in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) Presenilin-1 C-Terminal Fragment, rhPresenilin-2 C-Terminal Fragment, or rhPresenilin-2 N-Terminal Fragment is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 121130 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AD3EC 3.4.23.-, Alzheimer disease 3, EC 3.4.23, FAD, presenilin 1, Presenilin1, Protein S182, PS-1, PS1presenilin-1, PSEN1, PSNL1, S182 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5663 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction