missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Plexin B1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
£168.00 - £432.00
Specifications
| Antigen | Plexin B1 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18715187
![]() |
R&D Systems
AF3749-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18039631
![]() |
R&D Systems
AF3749 |
100 μg |
£432.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Plexin B1 Polyclonal specifically detects Plexin B1 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| Plexin B1 | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| plexin B1, PLXN5, PLXNB1, SEP, Transmembrane Protein SEP | |
| PLXNB1 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| O43157 | |
| 5364 | |
| E. coli-derived recombinant human Plexin B1 Leu35-Val150 Accession # O43157 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human Plexin B1 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human Plexin B3 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title