missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Plexin B1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
Brand: R&D Systems AF3749
This item is not returnable.
View return policy
Description
Plexin B1 Polyclonal specifically detects Plexin B1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Plexin B1 | |
| Polyclonal | |
| Unconjugated | |
| O43157 | |
| PLXNB1 | |
| E. coli-derived recombinant human Plexin B1 Leu35-Val150 Accession # O43157 | |
| 100 μg | |
| Primary | |
| Detects human Plexin B1 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human Plexin B3 is observed. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| plexin B1, PLXN5, PLXNB1, SEP, Transmembrane Protein SEP | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5364 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction