missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Pleiotrophin/PTN Antibody, R&D Systems™

Rat Monoclonal Antibody
£205.00 - £1730.00
Specifications
| Antigen | Pleiotrophin/PTN |
|---|---|
| Clone | 851406R |
| Dilution | Immunocytochemistry 8-25 ug/mL |
| Applications | Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18784988
![]() |
R&D Systems
MAB252R-SP |
25 μg |
£205.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18767529
![]() |
R&D Systems
MAB252R-100 |
100 μg |
£438.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18108820
|
R&D Systems
MAB252R-500 |
500 ug |
£1730.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18172501
|
R&D Systems
MAB252R-01M |
1 mg |
£1460.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Pleiotrophin/PTN Monoclonal specifically detects Pleiotrophin/PTN in Human samples. It is validated for ELISA, Immunocytochemistry.Specifications
| Pleiotrophin/PTN | |
| Immunocytochemistry 8-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| P21246 | |
| 5764 | |
| IgG2b | |
| Protein A or G purified from cell culture supernatant | |
| Detects human Pleiotrophin in direct ELISAs. |
| 851406R | |
| Immunocytochemistry | |
| Unconjugated | |
| Rat | |
| Human | |
| HARP, HBBM, HB-GAM, HBGF8, HBNF, HBNF1, HBNF-1, Heparin-binding brain mitogen, HNGF-8, NEGF1HBGF-8, neurite growth-promoting factor 1, neurite growth-promoting factor1), OSF-1, Osteoblast-specific factor 1, pleiotrophin, PTN | |
| PTN | |
| Primary | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title