missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Pleiotrophin/PTN Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB252R-500
This item is not returnable.
View return policy
Description
Pleiotrophin/PTN Monoclonal specifically detects Pleiotrophin/PTN in Human samples. It is validated for ELISA, Immunocytochemistry.
Specifications
| Pleiotrophin/PTN | |
| Monoclonal | |
| Unconjugated | |
| Supplied as a solution in PBS. *Small pack size (SP) is supplied as a 0.2 um filtered solution in PBS. with No Preservative | |
| HARP, HBBM, HB-GAM, HBGF8, HBNF, HBNF1, HBNF-1, Heparin-binding brain mitogen, HNGF-8, NEGF1HBGF-8, neurite growth-promoting factor 1, neurite growth-promoting factor1), OSF-1, Osteoblast-specific factor 1, pleiotrophin, PTN | |
| Rat | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 5764 | |
| Human | |
| Purified |
| Immunocytochemistry | |
| 851406R | |
| Immunocytochemistry 8-25 ug/mL | |
| P21246 | |
| PTN | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Pleiotrophin/PTN Gly33-Asp168 Accession No. P21246 | |
| 500 ug | |
| Primary | |
| Detects human Pleiotrophin in direct ELISAs. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction