missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Phospho-STAT5a/b (Y694/Y699) Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB4190-SP
This item is not returnable.
View return policy
Description
STAT5a/b Monoclonal specifically detects STAT5a/b in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| STAT5a/b | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| MGF, STAT5, STAT5A signal transducer and activator of transcription 5A | |
| Phosphopeptide containing human STAT5b Y699 site (amino acid sequence of this peptide is identical to a corresponding region of human STAT5a containing Y694). Accession # P51692 | |
| 25 μg | |
| Primary | |
| Detects human STAT5a/b when phosphorylated at Y694/Y699 in Western blots. | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 941212 | |
| Western Blot 2 ug/mL, Immunocytochemistry 0.3-25 ug/mL | |
| P51692 | |
| Rat | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 6776 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction