missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PGLYRP1/PGRP-S Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
£180.00 - £517.00
Specifications
| Antigen | PGLYRP1/PGRP-S |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18715787
![]() |
R&D Systems
AF2590-SP |
25 μg |
£180.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18031902
|
R&D Systems
AF2590 |
100 μg |
£517.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PGLYRP1/PGRP-S Polyclonal specifically detects PGLYRP1/PGRP-S in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
| PGLYRP1/PGRP-S | |
| Western Blot 0.1 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| peptidoglycan recognition protein 1, Peptidoglycan recognition protein short, PGLYRP, PGRPpeptidoglycan recognition protein, PGRPS, PGRP-S, PGRP-SMGC126894, PGRPSMGC126896, TAG7, TNF superfamily, member 3 (LTB)-like (peptidoglycan recognition protein), TNFSF3L | |
| PGLYRP1 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, ELISA | |
| Unconjugated | |
| RUO | |
| O75594 | |
| 8993 | |
| Mouse myeloma cell line NS0-derived recombinant human PGLYRP1/PGRP-S Gln22-Pro196 Accession # O75594 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human PGLYRP1/PGRP-S in ELISAs and Western blots. In sandwich immunoassays, less than 0.05% cross-reactivity with recombinant human PGRP-1 beta and recombinant mouse PGRP-S is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title