missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PGLYRP1/PGRP-S Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF2590-SP
This item is not returnable.
View return policy
Description
PGLYRP1/PGRP-S Polyclonal specifically detects PGLYRP1/PGRP-S in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).
Specifications
| PGLYRP1/PGRP-S | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| peptidoglycan recognition protein 1, Peptidoglycan recognition protein short, PGLYRP, PGRPpeptidoglycan recognition protein, PGRPS, PGRP-S, PGRP-SMGC126894, PGRPSMGC126896, TAG7, TNF superfamily, member 3 (LTB)-like (peptidoglycan recognition protein), TNFSF3L | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 8993 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
| O75594 | |
| PGLYRP1 | |
| Mouse myeloma cell line NS0-derived recombinant human PGLYRP1/PGRP-S Gln22-Pro196 Accession # O75594 | |
| 25 μg | |
| Primary | |
| Detects human PGLYRP1/PGRP-S in ELISAs and Western blots. In sandwich immunoassays, less than 0.05% cross-reactivity with recombinant human PGRP-1 beta and recombinant mouse PGRP-S is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction