missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PDGF-AA Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB221-100
This item is not returnable.
View return policy
Description
PDGF-AA Monoclonal specifically detects PDGF-AA in Human samples. It is validated for Neutralization.
Specifications
| PDGF-AA | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| PDGF1, PDGF-A, PDGFAA | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5154 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Neutralization | |
| 114506 | |
| Neutralization 0.2-1.0 ug/mL | |
| P04085 | |
| PDGFA | |
| E. coli-derived recombinant human PDGF-AA Ser87-Thr211 Accession # P04085 | |
| 100 μg | |
| Primary | |
| Detects human PDGF-AA in direct ELISAs. In direct ELISAs, approximately 20% cross-reactivity with recombinant human (rh) PDGF-AB is observed and no cross-reactivity with rhPDGF-BB, rhPlGF, rhVEGF206, rhVEGF B 186 rhVEGF C, rhVEGF D, or rhCTGF is observed. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction