missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PDGF-AA Antibody, R&D Systems™

Mouse Monoclonal Antibody
£168.00 - £1550.00
Specifications
| Antigen | PDGF-AA |
|---|---|
| Clone | 114506 |
| Dilution | Neutralization 0.2-1.0 ug/mL |
| Applications | Neutralization |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18794557
![]() |
R&D Systems
MAB221-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18748968
![]() |
R&D Systems
MAB221-100 |
100 μg |
£386.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18721199
![]() |
R&D Systems
MAB221-500 |
500 μg |
£1550.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PDGF-AA Monoclonal specifically detects PDGF-AA in Human samples. It is validated for Neutralization.Specifications
| PDGF-AA | |
| Neutralization 0.2-1.0 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| P04085 | |
| 5154 | |
| E. coli-derived recombinant human PDGF-AA Ser87-Thr211 Accession # P04085 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human PDGF-AA in direct ELISAs. In direct ELISAs, approximately 20% cross-reactivity with recombinant human (rh) PDGF-AB is observed and no cross-reactivity with rhPDGF-BB, rhPlGF, rhVEGF206, rhVEGF B 186 rhVEGF C, rhVEGF D, or rhCTGF is observed. |
| 114506 | |
| Neutralization | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| PDGF1, PDGF-A, PDGFAA | |
| PDGFA | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title