missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PDGF-AA Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB2211-SP
This item is not returnable.
View return policy
Description
PDGF-AA Monoclonal specifically detects PDGF-AA in Human samples. It is validated for Western Blot.
Specifications
| PDGF-AA | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| PDGF1, PDGF-A, PDGFAA | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5154 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot | |
| 114503 | |
| Western Blot 1 ug/mL | |
| P04085 | |
| PDGFA | |
| E. coli-derived recombinant human PDGF-AA Ser87-Thr211 Accession # P04085 | |
| 25 μg | |
| Primary | |
| Detects human PDGF-AA in direct ELISAs and Western blots. In direct ELISAs, this antibody shows 25% cross-reactivity with recombinant human (rh) PDGF-AB and no cross-reactivity with rhPDGF-BB, rhPlGF, rhVEGF, rhVEGF-C, rhVEGF-D, rhCTGF, or rhLDGF. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction