missing translation for 'onlineSavingsMsg'
Learn More

Human PDGF-AA Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

£168.00 - £492.00

Specifications

Antigen PDGF-AA
Clone 114503
Dilution Western Blot 1 ug/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18790927
GREENER_CHOICE
View Documents
R&D Systems
MAB2211-SP
25 μg
£168.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18113181
View Documents
R&D Systems
MAB2211
500 μg
£492.00
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PDGF-AA Monoclonal specifically detects PDGF-AA in Human samples. It is validated for Western Blot.
Specifications

Specifications

PDGF-AA
Western Blot 1 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
PDGF1, PDGF-A, PDGFAA
PDGFA
IgG2b
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
114503
Western Blot
Unconjugated
Mouse
Human
P04085
5154
E. coli-derived recombinant human PDGF-AA Ser87-Thr211 Accession # P04085
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human PDGF-AA in direct ELISAs and Western blots. In direct ELISAs, this antibody shows 25% cross-reactivity with recombinant human (rh) PDGF-AB and no cross-reactivity with rhPDGF-BB, rhPlGF, rhVEGF, rhVEGF-C, rhVEGF-D, rhCTGF, or rhLDGF.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.