missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human p62/SQSTM1 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
£171.15 - £459.90
Specifications
| Antigen | p62/SQSTM1 |
|---|---|
| Clone | 2533B |
| Dilution | Western Blot 1 ug/mL, Simple Western 20 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunoprecipitation 1 ug/1 mg cell lysate, Immunocytochemistry 8-25 ug/mL, Knockout Validated |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry, Immunoassay |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18732957
![]() |
R&D Systems
MAB80281-SP |
25 μg |
£181.00 £171.15 / 25µg Save £9.85 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18751297
![]() |
R&D Systems
MAB80281-100 |
100 μg |
£487.00 £459.90 / 100µg Save £27.10 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
p62/SQSTM1 Monoclonal specifically detects p62/SQSTM1 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry, Knockout Validated.Specifications
| p62/SQSTM1 | |
| Western Blot 1 ug/mL, Simple Western 20 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunoprecipitation 1 ug/1 mg cell lysate, Immunocytochemistry 8-25 ug/mL, Knockout Validated | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| A170, Autophagy Receptor P62, DMRV, EBI3-associated protein of 60 kDa, EBI3-associated protein p60, EBIAP, FTDALS3, NADGP, ORCA, OSIL, oxidative stress induced like, p60PDB3, p62, p62B, Paget disease of bone 3, PDB3, phosphotyrosine independent ligand for the Lck SH2 domain p62, Phosphotyrosine-independent ligand for the Lck SH2 domain of 62 kDa, sequestosome 1, sequestosome-1, SQSTM1, Ubiquitin-binding protein p62, ZIP3 | |
| E. coli-derived recombinant human p62/SQSTM1, Asp368-Leu440, Accession # Q13501 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 2533B | |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry, Immunoassay | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q13501 | |
| 8878 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title