Learn More
Human p62/SQSTM1 Antibody, R&D Systems™
GREENER_CHOICE
Rabbit Monoclonal Antibody
Brand:  R&D Systems MAB80281-100
 View more versions of this product
Product Code. 18751297
  • ONLINE EXCLUSIVE PRICE
    £487.00
    £459.90 / 100µg
    Save £27.10
    5% Off
Description
p62/SQSTM1 Monoclonal specifically detects p62/SQSTM1 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry, Knockout Validated.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
p62/SQSTM1
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
A170, Autophagy Receptor P62, DMRV, EBI3-associated protein of 60 kDa, EBI3-associated protein p60, EBIAP, FTDALS3, NADGP, ORCA, OSIL, oxidative stress induced like, p60PDB3, p62, p62B, Paget disease of bone 3, PDB3, phosphotyrosine independent ligand for the Lck SH2 domain p62, Phosphotyrosine-independent ligand for the Lck SH2 domain of 62 kDa, sequestosome 1, sequestosome-1, SQSTM1, Ubiquitin-binding protein p62, ZIP3
E. coli-derived recombinant human p62/SQSTM1, Asp368-Leu440, Accession # Q13501
100 μg
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
IgG
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry, Immunoassay
2533B
Western Blot 1 ug/mL, Simple Western 20 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunoprecipitation 1 ug/1 mg cell lysate, Immunocytochemistry 8-25 ug/mL, Knockout Validated
Q13501
Rabbit
Protein A or G purified from cell culture supernatant
RUO
8878
Human
Purified