missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Osterix/Sp7 MAb (Clone 764746), R&D Systems™

Mouse Monoclonal Antibody
£156.00 - £402.00
Specifications
| Antigen | Osterix/Sp7 |
|---|---|
| Clone | 764746 |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18791477
![]() |
R&D Systems
MAB75471-SP |
25 μg |
£156.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18711187
![]() |
R&D Systems
MAB75471-100 |
100 μg |
£402.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Osterix/Sp7 Monoclonal antibody specifically detects Osterix/Sp7 in Human samples. It is validated for Western BlotSpecifications
| Osterix/Sp7 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| MGC126598, OSX, OSXosterix, Sp7, Sp7 transcription factor, transcription factor Sp7, Zinc finger protein osterix | |
| E. coli-derived recombinant human Osterix/Sp7, Met19-Leu288, Accession # Q8TDD2 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 764746 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q8TDD2 | |
| 121340 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title