Learn More
Human Osterix/Sp7 MAb (Clone 764746), R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB75471-100
 View more versions of this product
Product Code. 18711187
  • £402.00 / 100µg
Description
Osterix/Sp7 Monoclonal antibody specifically detects Osterix/Sp7 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Osterix/Sp7
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
MGC126598, OSX, OSXosterix, Sp7, Sp7 transcription factor, transcription factor Sp7, Zinc finger protein osterix
E. coli-derived recombinant human Osterix/Sp7, Met19-Leu288, Accession # Q8TDD2
100 μg
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
IgG2b
Western Blot
764746
Western Blot 1 ug/mL
Q8TDD2
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
121340
Human
Purified