missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Osteoprotegerin/TNFRSF11B Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 5 publications
£168.00 - £1550.00
Specifications
| Antigen | Osteoprotegerin/TNFRSF11B |
|---|---|
| Clone | 69127 |
| Dilution | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18702528
![]() |
R&D Systems
MAB8051-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18734489
![]() |
R&D Systems
MAB8051-100 |
100 μg |
£389.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18756018
![]() |
R&D Systems
MAB8051-500 |
500 μg |
£1550.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Osteoprotegerin/TNFRSF11B Monoclonal specifically detects Osteoprotegerin/TNFRSF11B in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).Spezifikation
| Osteoprotegerin/TNFRSF11B | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| O00300 | |
| 4982 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Osteoprotegerin/TNFRSF11B Glu22-Leu401 Accession # O00300 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Osteoprotegerin/TNFRSF11B in ELISAs. It has been reported (1), and R&D Systems testing has confirmed, that this antibody shows strong cross-reactivity with an unrelated protein, human Carbonic Anhydrase II. In Western blots, this antibody does not cross-react with recombinant human (rh) 4-1BB, rhCD27, rhCD30, recombinant mouse (rm) CD30, rmCD40, rhDR3, rhDR6, rhFas, rmFas, rhGITR, rmGITR, rhHVEM, rhLT beta R, rhNGF R, rmOPG, rhTNF RI, rmTNF RI, rhTNF RII, or rmTNF RII. |
| 69127 | |
| ELISA | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| OCIF, OCIFMGC29565, OPG, OPGtumor necrosis factor receptor superfamily member 11B, Osteoclastogenesis inhibitory factor, Osteoprotegerin, TNFRSF11B, TR1, tumor necrosis factor receptor superfamily, member 11b | |
| TNFRSF11B | |
| IgG2a | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts