missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Osteoprotegerin/TNFRSF11B Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 5 publications
Brand: R&D Systems MAB8051-SP
This item is not returnable.
View return policy
Description
Osteoprotegerin/TNFRSF11B Monoclonal specifically detects Osteoprotegerin/TNFRSF11B in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).
Specifications
| Osteoprotegerin/TNFRSF11B | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| OCIF, OCIFMGC29565, OPG, OPGtumor necrosis factor receptor superfamily member 11B, Osteoclastogenesis inhibitory factor, Osteoprotegerin, TNFRSF11B, TR1, tumor necrosis factor receptor superfamily, member 11b | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 4982 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| ELISA | |
| 69127 | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| O00300 | |
| TNFRSF11B | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Osteoprotegerin/TNFRSF11B Glu22-Leu401 Accession # O00300 | |
| 25 μg | |
| Primary | |
| Detects human Osteoprotegerin/TNFRSF11B in ELISAs. It has been reported (1), and R&D Systems testing has confirmed, that this antibody shows strong cross-reactivity with an unrelated protein, human Carbonic Anhydrase II. In Western blots, this antibody does not cross-react with recombinant human (rh) 4-1BB, rhCD27, rhCD30, recombinant mouse (rm) CD30, rmCD40, rhDR3, rhDR6, rhFas, rmFas, rhGITR, rmGITR, rhHVEM, rhLT beta R, rhNGF R, rmOPG, rhTNF RI, rmTNF RI, rhTNF RII, or rmTNF RII. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction