missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Osteocrin Antibody, R&D Systems™

Rat Monoclonal Antibody
£156.00 - £346.00
Specifications
| Antigen | Osteocrin |
|---|---|
| Clone | 311334 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18729617
![]() |
R&D Systems
MAB9669-SP |
25 μg |
£156.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18744588
![]() |
R&D Systems
MAB9669-100 |
100 μg |
£346.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Osteocrin Monoclonal specifically detects Osteocrin in Human samples. It is validated for ELISA.Specifications
| Osteocrin | |
| ELISA | |
| Monoclonal | |
| Supernatant | |
| Human | |
| P61366 | |
| 344901 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 311334 | |
| ELISA | |
| Unconjugated | |
| Rat | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| MUSCLIN, osteocrin, OSTN | |
| E. coli-derived recombinant human Osteocrin Val28-Gly133 Accession # P61366 | |
| Primary | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Detects human Osteocrin in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title