missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Osteocrin Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB9669-100
This item is not returnable.
View return policy
Description
Osteocrin Monoclonal specifically detects Osteocrin in Human samples. It is validated for ELISA.
Specifications
| Osteocrin | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| MUSCLIN, osteocrin, OSTN | |
| E. coli-derived recombinant human Osteocrin Val28-Gly133 Accession # P61366 | |
| 100 μg | |
| 344901 | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| ELISA | |
| 311334 | |
| ELISA | |
| P61366 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human Osteocrin in direct ELISAs. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction