missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NKp80/KLRF1 C-Terminus Antibody, R&D Systems™

Sheep Polyclonal Antibody
£167.00 - £432.00
Specifications
| Antigen | NKp80/KLRF1 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells |
| Applications | Western Blot, Flow Cytometry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18746667
![]() |
R&D Systems
AF1900-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18051232
|
R&D Systems
AF1900 |
100 μg |
£432.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NKp80/KLRF1 Polyclonal specifically detects NKp80/KLRF1 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
| NKp80/KLRF1 | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells | |
| Polyclonal | |
| Sheep | |
| Human | |
| Activating coreceptor NKp80, CLEC5C, CLEC5CMGC119908, C-type lectin domain family 5 member C, killer cell lectin-like receptor F1, killer cell lectin-like receptor subfamily F member 1, killer cell lectin-like receptor subfamily F, member 1, KLRF1, Lectin-like receptor F1, MGC119907, MGC119909, ML, NKp80 | |
| KLRF1 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Flow Cytometry | |
| Unconjugated | |
| RUO | |
| Q9NZS2 | |
| 51348 | |
| Mouse myeloma cell line NS0-derived recombinant human NKp80/KLRF1 C-Terminus Val66-Tyr231 Accession # Q9NZS2 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human NKp80/KLRF1 C-Terminus in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) NKp44 and rhNKp30 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title