missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NKp80/KLRF1 C-Terminus Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF1900-SP
This item is not returnable.
View return policy
Description
NKp80/KLRF1 Polyclonal specifically detects NKp80/KLRF1 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| NKp80/KLRF1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Activating coreceptor NKp80, CLEC5C, CLEC5CMGC119908, C-type lectin domain family 5 member C, killer cell lectin-like receptor F1, killer cell lectin-like receptor subfamily F member 1, killer cell lectin-like receptor subfamily F, member 1, KLRF1, Lectin-like receptor F1, MGC119907, MGC119909, ML, NKp80 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 51348 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Flow Cytometry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells | |
| Q9NZS2 | |
| KLRF1 | |
| Mouse myeloma cell line NS0-derived recombinant human NKp80/KLRF1 C-Terminus Val66-Tyr231 Accession # Q9NZS2 | |
| 25 μg | |
| Primary | |
| Detects human NKp80/KLRF1 C-Terminus in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) NKp44 and rhNKp30 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction