missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Neuronal Pentraxin R/NPTXR Antibody, R&D Systems™

Sheep Polyclonal Antibody has been used in 3 publications
£185.00 - £483.00
Specifications
| Antigen | Neuronal Pentraxin R/NPTXR |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 μg/mL, Simple Western 20 μg/mL, Immunohistochemistry 5-15 μg/mL |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18707597
![]() |
R&D Systems
AF4414-SP |
25 μg |
£185.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18070242
|
R&D Systems
AF4414 |
100 μg |
£483.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Neuronal Pentraxin R/NPTXR Polyclonal specifically detects Neuronal Pentraxin R/NPTXR in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.Specifications
| Neuronal Pentraxin R/NPTXR | |
| Western Blot 0.1 μg/mL, Simple Western 20 μg/mL, Immunohistochemistry 5-15 μg/mL | |
| Polyclonal | |
| Sheep | |
| Human | |
| neuronal pentraxin receptor, NPR, NPTXR | |
| NPTXR | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| O95502 | |
| 23467 | |
| Mouse myeloma cell line NS0-derived recombinant human Pentraxin R/NPTXR Ala24-Lys499 Accession # O95502 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human Neuronal Pentraxin R/NPTXR in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) NPTX2 and rhNPTX1 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title