missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Neuronal Pentraxin R/NPTXR Antibody, R&D Systems™

Sheep Polyclonal Antibody has been used in 3 publications
Brand: R&D Systems AF4414-SP
This item is not returnable.
View return policy
Description
Neuronal Pentraxin R/NPTXR Polyclonal specifically detects Neuronal Pentraxin R/NPTXR in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.
Specifications
| Neuronal Pentraxin R/NPTXR | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| neuronal pentraxin receptor, NPR, NPTXR | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 23467 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 μg/mL, Simple Western 20 μg/mL, Immunohistochemistry 5-15 μg/mL | |
| O95502 | |
| NPTXR | |
| Mouse myeloma cell line NS0-derived recombinant human Pentraxin R/NPTXR Ala24-Lys499 Accession # O95502 | |
| 25 μg | |
| Primary | |
| Detects human Neuronal Pentraxin R/NPTXR in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) NPTX2 and rhNPTX1 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction