missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MYH7 Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Brand: R&D Systems MAB90961-100
This item is not returnable.
View return policy
Description
MYH7 Monoclonal specifically detects MYH7 in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| MYH7 | |
| Monoclonal | |
| Unconjugated | |
| Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
| cardiac muscle beta isoform, CMD1S, CMD1SMGC138376, CMH1, DKFZp451F047, MGC138378, MPD1, MYHCB, MyHC-beta, myhc-slow, myopathy, distal 1, myosin heavy chain (AA 1-96), myosin, heavy chain 7, cardiac muscle, beta, myosin, heavy polypeptide 7, cardiac muscle, beta, myosin-7, rhabdomyosarcoma antigen MU-RMS-40.7A, SPMD, SPMM | |
| Human MYH7 synthetic peptide Accession # P12883 | |
| 100 μg | |
| Primary | |
| Detects human MYH7 in direct ELISAs and Western blots. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
| Western Blot, Immunocytochemistry | |
| 2021A | |
| Western Blot 0.5 ug/mL, Immunocytochemistry 5-25 ug/mL | |
| P12883 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 4625 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction