missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MYH7 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
£181.00 - £440.00
Specifications
| Antigen | MYH7 |
|---|---|
| Clone | 2021A |
| Dilution | Western Blot 0.5 ug/mL, Immunocytochemistry 5-25 ug/mL |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18784889
![]() |
R&D Systems
MAB90961-SP |
25 μg |
£181.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18101821
|
R&D Systems
MAB90961-100 |
100 μg |
£440.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MYH7 Monoclonal specifically detects MYH7 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
| MYH7 | |
| Western Blot 0.5 ug/mL, Immunocytochemistry 5-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
| cardiac muscle beta isoform, CMD1S, CMD1SMGC138376, CMH1, DKFZp451F047, MGC138378, MPD1, MYHCB, MyHC-beta, myhc-slow, myopathy, distal 1, myosin heavy chain (AA 1-96), myosin, heavy chain 7, cardiac muscle, beta, myosin, heavy polypeptide 7, cardiac muscle, beta, myosin-7, rhabdomyosarcoma antigen MU-RMS-40.7A, SPMD, SPMM | |
| Human MYH7 synthetic peptide Accession # P12883 | |
| Primary | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 2021A | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P12883 | |
| 4625 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Detects human MYH7 in direct ELISAs and Western blots. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title