missing translation for 'onlineSavingsMsg'
Learn More

Human MSPR/Ron Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

£168.00 - £1525.00

Specifications

Antigen MSPR/Ron
Clone 99211
Dilution Blockade of Receptor-ligand Interaction
Applications Blockade of Receptor-ligand Interaction
Classification Monoclonal
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18759028
GREENER_CHOICE
View Documents
R&D Systems
MAB691-SP
25 μg
£168.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18787578
GREENER_CHOICE
View Documents
R&D Systems
MAB691-100
100 μg
£379.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18760827
GREENER_CHOICE
View Documents
R&D Systems
MAB691-500
500 μg
£1525.00
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MSPR/Ron Monoclonal specifically detects MSPR/Ron in Human samples. It is validated for Blockade of Receptor-ligand Interaction.
Specifications

Specifications

MSPR/Ron
Blockade of Receptor-ligand Interaction
Monoclonal
Mouse
Human
Q04912
4486
Mouse myeloma cell line NS0-derived recombinant human MSP R Met1-Ser956 Accession # Q04912
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human MSP in direct ELISAs. In this format, this antibody shows less than 2% cross-reactivity with recombinant mouse (rm) MSP R. Recognizes an epitope found in the beta chain.
99211
Blockade of Receptor-ligand Interaction
Unconjugated
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
CD136, CD136 antigen, CDw136c-met-related tyrosine kinase, EC 2.7.10, EC 2.7.10.1, macrophage stimulating 1 receptor (c-met-related tyrosine kinase), MSP R, MSP receptor, MST1R, p185-Ron, Protein-tyrosine kinase 8, PTK8, PTK8 protein tyrosine kinase 8, Ron, RONmacrophage-stimulating protein receptor, soluble RON variant 1, soluble RON variant 2, soluble RON variant 3, soluble RON variant 4
MST1R
IgG1
Protein A or G purified from ascites
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.