missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MSPR/Ron Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB691-SP
This item is not returnable.
View return policy
Description
MSPR/Ron Monoclonal specifically detects MSPR/Ron in Human samples. It is validated for Blockade of Receptor-ligand Interaction.
Specifications
| MSPR/Ron | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD136, CD136 antigen, CDw136c-met-related tyrosine kinase, EC 2.7.10, EC 2.7.10.1, macrophage stimulating 1 receptor (c-met-related tyrosine kinase), MSP R, MSP receptor, MST1R, p185-Ron, Protein-tyrosine kinase 8, PTK8, PTK8 protein tyrosine kinase 8, Ron, RONmacrophage-stimulating protein receptor, soluble RON variant 1, soluble RON variant 2, soluble RON variant 3, soluble RON variant 4 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 4486 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Blockade of Receptor-ligand Interaction | |
| 99211 | |
| Blockade of Receptor-ligand Interaction | |
| Q04912 | |
| MST1R | |
| Mouse myeloma cell line NS0-derived recombinant human MSP R Met1-Ser956 Accession # Q04912 | |
| 25 μg | |
| Primary | |
| Detects human MSP in direct ELISAs. In this format, this antibody shows less than 2% cross-reactivity with recombinant mouse (rm) MSP R. Recognizes an epitope found in the beta chain. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction