missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse TGF-beta RI/ALK-5 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 5 publications
£187.00 - £411.00
Specifications
| Antigen | TGF-beta RI/ALK-5 |
|---|---|
| Clone | 141231 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18705768
![]() |
R&D Systems
MAB5871-SP |
25 μg |
£187.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18061952
|
R&D Systems
MAB5871 |
100 μg |
£411.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TGF-beta RI/ALK-5 Monoclonal specifically detects TGF-beta RI/ALK-5 in Human, Mouse samples. It is validated for Western Blot.Specifications
| TGF-beta RI/ALK-5 | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Human, Mouse | |
| BAA05023 | |
| 7046 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse TGF-beta RI/ALK-5 Ala21-Glu121 Accession # BAA05023 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human and mouse TGF-beta RI/ALK-5 in Western blots. In direct ELISAs and Western blots, this antibody shows no cross-reactivity with rrMIS RII, rhTGF-beta RII, rhTGF-beta RIIb, or rhTGF-beta RIII. |
| 141231 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AAT5, activin A receptor type II-like kinase, 53kD, activin A receptor type II-like kinase, 53kDa, Activin receptor-like kinase 5, ACVRLK4, ALK-5, ALK-5ALK5, EC 2.7.11, EC 2.7.11.30, LDS1A, LDS2A, Serine/threonine-protein kinase receptor R4, SKR4, TbetaR-I, TGFB1R1, TGF-beta receptor type I, TGF-beta receptor type-1, TGF-beta type I receptor, TGFbetaRI, TGFBR1, TGF-bRI, TGFR-1, transforming growth factor beta receptor I, transforming growth factor, beta receptor 1, transforming growth factor, beta receptor I (activin A receptor type II-likekinase, 53kD), Transforming growth factor-beta receptor type I | |
| TGFBR1 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title