missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse TGF-beta RI/ALK-5 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 5 publications
Brand: R&D Systems MAB5871-SP
This item is not returnable.
View return policy
Description
TGF-beta RI/ALK-5 Monoclonal specifically detects TGF-beta RI/ALK-5 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| TGF-beta RI/ALK-5 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| BAA05023 | |
| TGFBR1 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse TGF-beta RI/ALK-5 Ala21-Glu121 Accession # BAA05023 | |
| 25 μg | |
| Primary | |
| Detects human and mouse TGF-beta RI/ALK-5 in Western blots. In direct ELISAs and Western blots, this antibody shows no cross-reactivity with rrMIS RII, rhTGF-beta RII, rhTGF-beta RIIb, or rhTGF-beta RIII. | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| 141231 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AAT5, activin A receptor type II-like kinase, 53kD, activin A receptor type II-like kinase, 53kDa, Activin receptor-like kinase 5, ACVRLK4, ALK-5, ALK-5ALK5, EC 2.7.11, EC 2.7.11.30, LDS1A, LDS2A, Serine/threonine-protein kinase receptor R4, SKR4, TbetaR-I, TGFB1R1, TGF-beta receptor type I, TGF-beta receptor type-1, TGF-beta type I receptor, TGFbetaRI, TGFBR1, TGF-bRI, TGFR-1, transforming growth factor beta receptor I, transforming growth factor, beta receptor 1, transforming growth factor, beta receptor I (activin A receptor type II-likekinase, 53kD), Transforming growth factor-beta receptor type I | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7046 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction