missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Sulfatase Modifying Factor 1/SUMF1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£167.00 - £377.00
Specifications
| Antigen | Sulfatase Modifying Factor 1/SUMF1 |
|---|---|
| Clone | 329005 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18762568
![]() |
R&D Systems
MAB2779-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18111391
![]() |
R&D Systems
MAB2779 |
100 μg |
£377.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sulfatase Modifying Factor 1/SUMF1 Monoclonal specifically detects Sulfatase Modifying Factor 1/SUMF1 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| Sulfatase Modifying Factor 1/SUMF1 | |
| LYOPH | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| Q8NBK3.3 | |
| 285362 | |
| Mouse myeloma cell line NS0-derived recombinant human Sulfatase Modifying Factor 1/SUMF1 isoform 1 Ser34-Asp374 (Ser63Asn) Accession # Q8NBK3.3 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects recombinant human or mouse Sulfatase Modifying Factor 1/SUMF1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human SUMF2 or recombinant mouse SUMF2 is observed. |
| 329005 | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AAPA3037, EC 1.8.99.-, FGE1, FGEC-alpha-formylglycine-generating enzyme 1, FGly-generating enzyme, MGC131853, MGC150436, sulfatase modifying factor 1, sulfatase-modifying factor 1, UNQ3037 | |
| SUMF1 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title