missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Sulfatase Modifying Factor 1/SUMF1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB2779-SP
This item is not returnable.
View return policy
Description
Sulfatase Modifying Factor 1/SUMF1 Monoclonal specifically detects Sulfatase Modifying Factor 1/SUMF1 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Sulfatase Modifying Factor 1/SUMF1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Q8NBK3.3 | |
| SUMF1 | |
| Mouse myeloma cell line NS0-derived recombinant human Sulfatase Modifying Factor 1/SUMF1 isoform 1 Ser34-Asp374 (Ser63Asn) Accession # Q8NBK3.3 | |
| 25 μg | |
| Primary | |
| Detects recombinant human or mouse Sulfatase Modifying Factor 1/SUMF1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human SUMF2 or recombinant mouse SUMF2 is observed. | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 329005 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AAPA3037, EC 1.8.99.-, FGE1, FGEC-alpha-formylglycine-generating enzyme 1, FGly-generating enzyme, MGC131853, MGC150436, sulfatase modifying factor 1, sulfatase-modifying factor 1, UNQ3037 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 285362 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction