missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse SMOC-2 Isoform 1 Antibody, R&D Systems™

Sheep Polyclonal Antibody has been used in 2 publications
£170.00 - £459.00
Specifications
| Antigen | SMOC-2 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18724937
![]() |
R&D Systems
AF5140-SP |
25 μg |
£170.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18091342
|
R&D Systems
AF5140 |
100 μg |
£459.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SMOC-2 Polyclonal specifically detects SMOC-2 in Human, Mouse samples. It is validated for Western Blot.Specifications
| SMOC-2 | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Sheep | |
| Human, Mouse | |
| bA270C4A.1, bA37D8.1, dJ421D16.1, MST117, MSTP117, MSTP140, SMAP2, SMAP-2, SMAP2Secreted modular calcium-binding protein 2, SMOC2, SMOC-2, smooth muscle associated protein 2, Smooth muscle-associated protein 2, SPARC related modular calcium binding 2, SPARC-related modular calcium-binding protein 2, thyroglobulin type-1 repeat containing protein | |
| SMOC2 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| AAH47583 | |
| 64094 | |
| Mouse myeloma cell line NS0-derived recombinant human SMOC-2 Isoform 1 Gln22-Gly446 Accession # AAH47583 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human and mouse SMOC-2 Isoform 1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human SMOC-1 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title