missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse SMOC-2 Isoform 1 Antibody, R&D Systems™
Sheep Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF5140
This item is not returnable.
View return policy
Description
SMOC-2 Polyclonal specifically detects SMOC-2 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| SMOC-2 | |
| Polyclonal | |
| Unconjugated | |
| AAH47583 | |
| SMOC2 | |
| Mouse myeloma cell line NS0-derived recombinant human SMOC-2 Isoform 1 Gln22-Gly446 Accession # AAH47583 | |
| 100 μg | |
| Primary | |
| Detects human and mouse SMOC-2 Isoform 1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human SMOC-1 is observed. | |
| Human, Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| bA270C4A.1, bA37D8.1, dJ421D16.1, MST117, MSTP117, MSTP140, SMAP2, SMAP-2, SMAP2Secreted modular calcium-binding protein 2, SMOC2, SMOC-2, smooth muscle associated protein 2, Smooth muscle-associated protein 2, SPARC related modular calcium binding 2, SPARC-related modular calcium-binding protein 2, thyroglobulin type-1 repeat containing protein | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 64094 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction