missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat LC3B Antibody, R&D Systems™

Rabbit Monoclonal Antibody
£179.00 - £451.00
Specifications
| Antigen | LC3B |
|---|---|
| Clone | 1251B |
| Dilution | Western Blot 0.1 ug/mL, Simple Western 5 ug/mL, Immunocytochemistry 1-25 ug/mL, Knockout Validated 0.1 ug/mL |
| Applications | Western Blot, Immunocytochemistry, KnockDown |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18731798
![]() |
R&D Systems
MAB85582-SP |
25 μg |
£179.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18736568
![]() |
R&D Systems
MAB85582-100 |
100 μg |
£451.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LC3B Monoclonal specifically detects LC3B in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunocytochemistry, Knockout Validated.Specifications
| LC3B | |
| Western Blot 0.1 ug/mL, Simple Western 5 ug/mL, Immunocytochemistry 1-25 ug/mL, Knockout Validated 0.1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Apg8b, ATG8F, Autophagy-related protein LC3 B, Autophagy-related ubiquitin-like modifier LC3 B, LC3B, lc3b autophagy marker, lc3-i, ii, LC3II, MAP1 light chain 3-like protein 2, MAP1A/1BLC3, MAP1A/MAP1B LC3 B, map1lc3b, MAP1LC3B-a, microtubule associated protein 1 light chain 3 beta, microtubule associated protein 3 b, microtubule associated protein 3 beta, microtubule-associated proteins 1A/1B light chain 3, microtubule-associated proteins 1A/1B light chain 3B | |
| Human LC3B synthetic peptide, Accession # Q9GZQ8 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human, mouse, and rat LC3B in Western blots. |
| 1251B | |
| Western Blot, Immunocytochemistry, KnockDown | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q9GZQ8 | |
| 81631 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
RUO
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title