missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat LC3B Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB85582-SP
This item is not returnable.
View return policy
Description
LC3B Monoclonal specifically detects LC3B in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunocytochemistry, Knockout Validated.
Specifications
| LC3B | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Apg8b, ATG8F, Autophagy-related protein LC3 B, Autophagy-related ubiquitin-like modifier LC3 B, LC3B, lc3b autophagy marker, lc3-i, ii, LC3II, MAP1 light chain 3-like protein 2, MAP1A/1BLC3, MAP1A/MAP1B LC3 B, map1lc3b, MAP1LC3B-a, microtubule associated protein 1 light chain 3 beta, microtubule associated protein 3 b, microtubule associated protein 3 beta, microtubule-associated proteins 1A/1B light chain 3, microtubule-associated proteins 1A/1B light chain 3B | |
| Human LC3B synthetic peptide, Accession # Q9GZQ8 | |
| 25 μg | |
| Primary | |
| Detects human, mouse, and rat LC3B in Western blots. | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunocytochemistry, KnockDown | |
| 1251B | |
| Western Blot 0.1 ug/mL, Simple Western 5 ug/mL, Immunocytochemistry 1-25 ug/mL, Knockout Validated 0.1 ug/mL | |
| Q9GZQ8 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 81631 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RUO
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering