missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat IGSF8/CD316 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
£168.00 - £381.00
Specifications
| Antigen | IGSF8/CD316 |
|---|---|
| Clone | 2587G |
| Dilution | Western Blot 2 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/106 cells, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18713007
![]() |
R&D Systems
MAB31171-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18760707
![]() |
R&D Systems
MAB31171-100 |
100 μg |
£381.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IGSF8/CD316 Monoclonal antibody specifically detects IGSF8/CD316 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
| IGSF8/CD316 | |
| Western Blot 2 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/106 cells, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CD316, CD316 antigen, CD81P3, CD81P3LIR-D1, EWI2, EWI-2, EWI2CD81 partner 3, Glu-Trp-Ile EWI motif-containing protein 2, IgSF8, immunoglobulin superfamily, member 8, KCT4, KCT-4, Keratinocytes-associated transmembrane protein 4, PGRL, PGRLimmunoglobulin superfamily member 8 | |
| Human embryonic kidney cell, HEK293-derived human IGSF8/CD316, Arg28-Thr579, Accession # Q969P0-1 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 2587G | |
| Western Blot, Flow Cytometry, CyTOF | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q969P0-1 | |
| 93185 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title